missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ VISTA/B7-H5/PD-1H Antibody (onvatilimab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27970-1mg
This item is not returnable.
View return policy
Description
VISTA/B7-H5/PD-1H Monoclonal antibody specifically detects VISTA/B7-H5/PD-1H in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| VISTA/B7-H5/PD-1H | |
| Monoclonal | |
| Unconjugated | |
| Q9H7M9 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| onvatilimab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR | |
| B7-H5/ VISTA | |
| 1 mg | |
| Cancer, Cardiovascular Biology, Immunology | |
| 64115 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction