missing translation for 'onlineSavingsMsg'
Learn More

VISTA/B7-H5/PD-1H Antibody (mam82), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18397981 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397981 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18397981 Supplier Novus Biologicals Supplier No. NBP311974JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

VISTA/B7-H5/PD-1H Monoclonal antibody specifically detects VISTA/B7-H5/PD-1H in Mouse samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen VISTA/B7-H5/PD-1H
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone mam82
Conjugate Janelia Fluor 646
Dilution ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR
Host Species Mouse
Immunogen The antibody was generated by immunising a VISTA/B7-H5/PD-1H-KO mouse with mouse VISTA/B7-H5/PD-1H Ig fusion protein and adjuvant.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 64115
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.