missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VISTA/B7-H5/PD-1H Antibody (730823), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB71263JF646
400.35 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
VISTA/B7-H5/PD-1H Monoclonal antibody specifically detects VISTA/B7-H5/PD-1H in Human samples. It is validated for Flow Cytometry
Specifications
| VISTA/B7-H5/PD-1H | |
| Monoclonal | |
| Janelia Fluor 646 | |
| B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR | |
| Partial recombinant human VISTA/B7-H5 (amino acids 33-194) [UniProt Q9H7M9] | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Immunology | |
| 64115 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| 730823 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction