Learn More
Macherey-Nagel™ Visocolor™ Eco Colorimetric and Titrimetric Test Kits
Run a wide range of water testing assays. Macherey Nagel™ Visocolor™ Eco Colorimetric and Titrimetric Test Kits avoid hazardous substances wherever possible.
Brand:  Macherey-Nagel™ 935079
Code : NEW
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 10216431
  • £33.30 / Each
Estimated Shipment: 29-07-2024
to see stock.
Add to basket
Description
Description
  • Colorimetric and titrimetric test kits
  • Avoids hazardous substances wherever possible
Specifications
Specifications
2 · 5 · 10 · 15 · 20 mg/l PO43-
Visocolor Alpha Phosphate
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • VISOCOLOR alpha Phosphate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : sulfuric acid, sodium disulfite