missing translation for 'onlineSavingsMsg'
Learn More

Mutexil™ Non-woven Blue High-filter Visitors Face Masks

Product Code. 11725473
Change view
Click to view available options
Color:
Blue
White
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11725473 White Pack of 1000
11765463 Blue Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11725473 Supplier Mutexil™ Supplier No. 4561016
Only null left
Add to basket
Add to basket

Use Mutexil™ Non-woven Blue High-filter Visitors Face Masks when visiting a manufacturer or to visiting patients.

  • Non-woven
  • Blue
  • Easily fastened
  • High filter for enhanced safety

Specifications

Product Type General Purpose Face Mask
Color White
Construction Non-woven
For Use With (Application) For used to visit a manufacture or to visit patients
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.