missing translation for 'onlineSavingsMsg'
Learn More
Learn More
visinin-like 1 Antibody (3A9), Novus Biologicals™
Mouse Monoclonal Antibody
£129.00 - £367.00
Specifications
| Antigen | visinin-like 1 |
|---|---|
| Clone | 3A9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:500-1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18184375
|
Novus Biologicals
NBP2-25167SS |
0.025 mL |
£129.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18197613
|
Novus Biologicals
NBP2-25167 |
0.1 mL |
£367.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
visinin-like 1 Monoclonal specifically detects visinin-like 1 in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| visinin-like 1 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine | |
| P62760 | |
| 7447 | |
| Full length human visinin-like 1 [UniProt# P62760] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 22 kDa |
| 3A9 | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:500-1:1000 | |
| Monoclonal | |
| Mouse | |
| Stem Cell Markers | |
| 50% PBS, 50% glycerol with 5mM Sodium Azide | |
| HLP3Hippocalcin-like protein 3, HPCAL3, HUVISL1, VILIP-1, VILIPVLP-1, visinin-like 1, visinin-like protein 1, VISL1 | |
| VSNL1 | |
| IgG1 | |
| Affinity Purified | |
| Human, Bovine, Mouse and Rat. Other species have not been tested. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title