missing translation for 'onlineSavingsMsg'
Learn More

visinin-like 1 Antibody (2D11), DyLight 594, Novus Biologicals™

Product Code. 18425245 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425245 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425245 Supplier Novus Biologicals Supplier No. NBP225166DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

visinin-like 1 Monoclonal antibody specifically detects visinin-like 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry Free-Floating
TRUSTED_SUSTAINABILITY

Specifications

Antigen visinin-like 1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Free Floating)
Classification Monoclonal
Clone 2D11
Conjugate DyLight 594
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry - Free Floating
Formulation 50mM Sodium Borate
Gene Alias HLP3Hippocalcin-like protein 3, HPCAL3, HUVISL1, VILIP-1, VILIPVLP-1, visinin-like 1, visinin-like protein 1, VISL1
Host Species Mouse
Immunogen Full length human visinin-like 1 [UniProt# P62760]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7447
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.