missing translation for 'onlineSavingsMsg'
Learn More

Viscosity standard, Specpure™, nominally 10cSt @ 40 and 2.7cSt @ 100

Product Code. 15417375
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 15417375

Brand: Thermo Scientific Alfa Aesar 047036.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 8042-47-5 | C16H10N2Na2O7S2 | 452.363 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 8042-47-5
Odor Odorless
Flash Point >110°C (230°F)
Physical Form Liquid
Molecular Formula C16H10N2Na2O7S2
MDL Number MFCD00084411
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
Molecular Weight (g/mol) 452.363
PubChem CID 9566064
Quantity 500 mL
Grade Specpure
Chemical Name or Material Viscosity standard, Nominally 10cSt at 40 and 2.7cSt at 100
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.