missing translation for 'onlineSavingsMsg'
Learn More

Viscosity Reference Standard - approximately 100 cSt at 40°C and approximately 16.8 cSt at 100°C., VHG™

Product Code. 17971006
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17971006 100 mL 100mL
17951856 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17971006 Supplier VHG Labs Supplier No. VHGVISC100A100
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Viscosity Reference Standard - approximately 100 cSt at 40°C and approximately 16.8 cSt at 100°C.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Viscosity
Methods Viscometer
Analyte or Component Names Viscosity
Shipping Condition Room Temperature
Concentration 100 cSt
Certifications/Compliance ISO 17034
Solvent or Matrix Hydrocarbons
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.