Learn More
Poulten Selfe™ Viscometer Holder
Product Code. 13161093
Click to view available options
Size:
Cannon-Fenske Size 25 to 500
Suspended Level Sizes 1 to 4
Suspended Level Sizes 4a to 5
U-tube Reverse Flow Sizes 1 to 9
U-tube Size A to F
Unit Size:
Each
Product Code. 13161093
Brand: Poulten Selfe™ 1818/A1
Only null left
Add to basket
Add to basket
Specifications
Product Type Viscometer Holder
For Use With (Equipment) 50 mm Bath Opening with Plastic Handle/U-tube Size A to F
Diameter (Metric) Opening 50 mm
Size U-tube Size A to F