Learn More
SI Analytics™ ViscoClock Plus
Product Code. 15679990 Shop All SI Analytics Products
Change view
Click to view available options
For Use With (Equipment):
Ubbelohde Viscometer
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
15679990 Ubbelohde Viscometer Each
1 options
Product Code. 15679990 Supplier SI Analytics™ Supplier No. 285417900
Only null left
Add to basket
Add to basket
ViscoClock plus is an electronic timing unit for Ubbelohde type glass capillary viscometers used to determine kinematic and relative viscosity.
  • Specially designed for Ubbelohde viscometers
  • Allows automatic measurement of flow times
  • Stand in high performance engineering PPA for temperatures up to 150°C
  • Convenient handling of Ubbelohde viscometer due to electromagnetic venting valve
  • Data storage including time, date, viscometer and sample identification number
TRUSTED_SUSTAINABILITY
Specifications
Product Type Electronic Timing Unit
For Use With (Equipment) Ubbelohde Viscometer