Learn More
Ansell™ Virtex™ 79-700 Series Blue Nitrile Immersion Gloves
Product Code. 10267842 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
50 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
10267842 11 50 pairs
11706703 10 50 pairs
10156072 7 50 pairs
11736703 8 50 pairs
11746703 9 50 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10267842 Supplier Ansell™ Supplier No. 79700110
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed specifically to offer outstanding comfort, flexibility and good chemical resistance. Ansell Virtex™ 79-700 Series Blue Nitrile Immersion Gloves provide all-day comfort in a lightweight glove.
  • Reusable
  • Anti-static according to EN1149
  • With Aquadri™-Ansell Moisture Management Technology™, which gives excellent moisture absorption capabilities and keeps users' hands drier for longer
  • Significantly lower tendency to shed than traditional cotton flock gloves
  • Ideal for light assembly in dry, fully wet or oily conditions, engine and system testing, paint and spray shops, food distribution
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nitrile
Cuff Material Nitrile
Product Type Immersion Gloves
Size 11
Style 79-700
Thickness (Metric) 0.2 mm
Glove Cuff Color Blue
Glove Cuff Style Straight
Liner Material Aquadri
Quantity 1 Pair
Resistant To Good chemical resistance
Series 79-700