Learn More
Virkon™ Virucidal Disinfectant
Product Code. 12348667
Change view
Click to view available options
Volume (Metric):
50 g
500 g
5 kg
Container Type:
Drum
Sachet
Squeeze Pack
Quantity:
1 Ea.
50/Pk.
6/Pk.
Unit Size:
5kg
Pack of 50
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Container Type Quantity unitSize
12348667 500 g Squeeze Pack 6/Pk. Pack of 6
12338667 50 g Sachet 50/Pk. Pack of 50
12358667 5 kg Drum 1 Ea. 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Use biocides safely. Always read the label and product information before use
Product Code. 12348667 Supplier Virkon™ Supplier No. 330002
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use biocides safely. Always read the label and product information before use
Clean and disinfect work surfaces and equipment in a single, one-step operation. Virkon™ Virucidal Disinfectant provides the widest proven spectrum of any disinfectant.
  • Use biocides safely. Always read the label and product information before use
  • Widest proven spectrum of any disinfectant - proven effective in independent tests against all 18 virus families affecting man and animals, including AIDS (HIV) and Hepatitis B
  • Excellent bactericide and fungicide - cleans and disinfects work surfaces and equipment in a single, one-step operation
  • Can be applied in powder form direct to spillages of blood and other body fluids
  • Easily dissolves to make a liquid disinfectant
  • Odourless, non-irritant to skin and eyes, non-toxic vapour phase, exceptionally low oral toxicity
  • Under COSHH requirements, no maximum exposure limit is required
  • Non-corrosive, non-bleaching and biodegradable
  • Any viral disease transmitted via surfaces or instruments contaminated with infected blood, body fluids, faeces or saliva can be controlled by correct application
  • For cleaning and disinfecting work surfaces and equipment
Specifications
Product Type Virucidal Disinfectant
Container Type Squeeze Pack
For Use With (Application) Virucidal Disinfectant
Quantity 6/Pk.
Volume (Metric) 500 g