missing translation for 'onlineSavingsMsg'
Learn More

Virilizer Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18410323 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410323 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410323 Supplier Novus Biologicals Supplier No. NBP121363AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Virilizer Polyclonal antibody specifically detects Virilizer in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Virilizer
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DKFZP434I116, DKFZp781B2117, fSAP121, functional spliceosome-associated protein 121, KIAA1429, MGC138493, MGC141940, protein virilizer homolog
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 200 and 250 of human Virilizer (KIAA1429) using the numbering given in entry NP_056311.2 (GeneID 25962).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 25962
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.