missing translation for 'onlineSavingsMsg'
Learn More

Virginiamycin M1 (~90%) (200 ug/mL in Methanol), TRC

Product Code. 30395762
Change view
Click to view available options
Quantity:
1x1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30395762 1x1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395762 Supplier Toronto Research Chemicals Supplier No. TRCKIT07581X1ML
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Virginiamycin M1
CAS 21411-53-0
Molecular Formula C28 H35 N3 O7
Quantity 1x1 mL
Synonym 3H-21,18-Nitrilo-1H,22H-pyrrolo[2,1-c][1,8,4,19]dioxadiazacyclotetracosine-1,7,16,22(4H,17H)-tetrone, 8,9,14,15,24,25-hexahydro-14-hydroxy-4,12-dimethyl-3-(1-methylethyl)-, (3R,4R,5E,10E,12E,14S)-, Mikamycin A (6CI,8CI), Ostreogrycin A (7CI), Virginiamycin M1, (3R,4R,5E,10E,12E,14S)-8,9,14,15,24,25-Hexahydro-14-hydroxy-4,12-dimethyl-3-(1-methylethyl)-3H-21,18-nitrilo-1H,22H-pyrrolo[2,1-c][1,8,4,19]dioxadiazacyclotetracosine-1,7,16,22(4H,17H)-tetrone, Antibiotic PA 114A, Antibiotic PA 114A1, Factor M, Factor M (antibiotic), NSC 244426, NSC 87432, PA 114A, Pristinamycin II, Pristinamycin IIA, RP 12536, Staphylomycin M1, Streptogramin A, Vernamycin A, [3R-(3R*,4R*,5E,10E,12E,14S*)]-8,9,14,15,24,25-Hexahydro-14-hydroxy-4,12-dimethyl-3-(1-methylethyl)-3H-21,18-nitrilo-1H,22H-pyrrolo[2,1-c][1,8,4,19]dioxadiazacyclotetracosine-1,7,16,22(4H,17H)-tetrone
InChI Formula InChI=1S/C28H35N3O7/c1-17(2)26-19(4)9-10-24(34)29-11-5-7-18(3)13-20(32)14-21(33)15-25-30-22(16-37-25)27(35)31-12-6-8-23(31)28(36)38-26/h5,7-10,13,16-17,19-20,26,32H,6,11-12,14-15H2,1-4H3,(H,29,34)/b7-5+,10-9+,18-13+/t19-,20-,26-/m1/s1
SMILES CC(C)[C@H]1OC(=O)C2=CCCN2C(=O)c3coc(CC(=O)C[C@H](O)\C=C(/C)\C=C\CNC(=O)\C=C\[C@H]1C)n3
Molecular Weight (g/mol) 525.59
Formula Weight 525.2475
Concentration or Composition (by Analyte or Components) 200 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.