missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral MIP-I Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB799-SP
This item is not returnable.
View return policy
Description
MIP-I Monoclonal specifically detects MIP-I in Viral samples. It is validated for Western Blot, Neutralization.
Specifications
| MIP-I | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Detects viral MIP-I in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant cytomegalovirus UL146, recombinant human (rh) CCL3, rhCCL22, recombinant mouse (rh) CCL3 or rmCCL22 is observed. | |
| Virus | |
| Purified |
| Western Blot, Neutralization | |
| 84420 | |
| Western Blot 1 ug/mL, Neutralization 0.1-0.5 ug/mL | |
| YP_001129366 | |
| E. coli-derived recombinant human herpes virus-8 MIP-I Ala25-Ala95 Accession # YP_001129366 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto