missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral HIV-1 Gag p24 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£185.00 - £552.00
Specifications
| Antigen | HIV-1 Gag p24 |
|---|---|
| Clone | 1042960 |
| Dilution | Western Blot 1 ug/mL, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18734217
![]() |
R&D Systems
MAB11188-SP |
25 μg |
£185.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18712007
|
R&D Systems
MAB11188-100 |
100 μg |
£552.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HIV-1 Gag p24 Monoclonal antibody specifically detects HIV-1 Gag p24 in Virus samples. It is validated for Western Blot, ELISASpecifications
| HIV-1 Gag p24 | |
| Western Blot 1 ug/mL, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 155030 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 1042960 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Virus | |
| Q77YG1_9HIV1 | |
| E. coli-derived recombinant viral HIV-1 Gag p24, Pro149-Gly354, Accession # Q77YG1_9HIV1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title