missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral HCMV IL-10 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems AF117-SP
This item is not returnable.
View return policy
Description
IL-10 Polyclonal specifically detects IL-10 in Viral samples. It is validated for Western Blot, Neutralization.
Specifications
| IL-10 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3586 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.15-0.9 ug/mL | |
| AAF63437 | |
| CeHV15gBCRF1 | |
| E. coli-derived recombinant viral HCMV IL-10 Ala26-Lys176 Accession # AAF63437 | |
| 25 μg | |
| Primary | |
| Detects viral HCMV IL-10 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse IL-10, recombinant canine IL-10, recombinant feline IL-10, recombinant cotton rat IL-10, recombinant rat IL-10, recombinant porcine IL-10, recombinant human IL-10, and recombinant Epstein-Barr virus encoded viral IL-10 is observed. | |
| Virus | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction