missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral EBV IL-10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB915-SP
This item is not returnable.
View return policy
Description
IL-10 Monoclonal specifically detects IL-10 in Viral samples. It is validated for Western Blot, Neutralization.
Specifications
| IL-10 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3586 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Neutralization | |
| 134216 | |
| Western Blot 1 ug/mL, Neutralization 0.1-0.4 ug/mL | |
| P03180 | |
| CeHV15gBCRF1 | |
| E. coli-derived recombinant viral EBV IL-10 Gln26-Arg170 Accession # P03180 | |
| 25 μg | |
| Primary | |
| Detects viral EBV IL-10 in direct ELISAs and Western blots. In Western blots, less than 10% cross-reactivity with recombinant canine IL-10, recombinant feline IL-10, recombinant mouse IL-10, recombinant porcine IL-10, and recombinant rat IL-10 is observed. | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction