Learn More
Invitrogen™ Viral CCI Monoclonal Antibody (112803)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA561746
Product Code. 30535680
  • £490.00 / 500µg
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
The family of T1/35 kDa proteins are secreted, soluble proteins encoded by the 35K virulence gene of many poxviruses. These proteins bind CC chemokines with high affinity and are now termed viral chemokine inhibitors (vCCI). Vaccinia virus (Strain Lister) vCCI shows greater than 90% amino acid sequence identity with vCCI from other orthopoxviruses and approximately 40% amino acid sequence identity with the leporipoxvirus T-1 proteins.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Viral CCI
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Mouse
Protein A/G
RUO
Virus
Antibody
IgG1
Neutralization, Western Blot
112803
Unconjugated
vCCI
S. frugiperda insect ovarian cell line Sf 21-derived recombinant viral CCI, Met17-Val258.
500 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized