missing translation for 'onlineSavingsMsg'
Learn More

Viperin Mouse anti-Human, Mouse, Clone: MaP.VIP, Novus Biologicals™

Product Code. 18330972 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18330972 100 μg 100µg
18611329 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18330972 Supplier Novus Biologicals Supplier No. NBP307918100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Viperin Monoclonal antibody specifically detects Viperin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Viperin
Applications Western Blot, Flow Cytometry, Immunoprecipitation, Immunofluorescence
Classification Monoclonal
Clone MaP.VIP
Concentration 0.1 mg/ml
Conjugate Unconjugated
Formulation PBS containing 0.1% BSA
Gene Alias 2510004L01Rik, cig33, cig5, Cytomegalovirus-induced gene 5 protein, interferon-inducible, radical S-adenosyl methionine domain containing 2, radical S-adenosyl methionine domain-containing protein 2, vig1, Viperin, Virus inhibitory protein, endoplasmic reticulum-associated
Host Species Mouse
Immunogen Recombinant mouse viperin fragment (residues 92-362)
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 91543
Target Species Human, Mouse
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.