missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viperin Mouse anti-Human, Mouse, Clone: MaP.VIP, Novus Biologicals™
Mouse Monoclonal Antibody
£396.00 - £684.00
Specifications
| Antigen | Viperin |
|---|---|
| Clone | MaP.VIP |
| Applications | Western Blot, Flow Cytometry, Immunoprecipitation, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18330972
|
Novus Biologicals
NBP3-07918-100UG |
100 μg |
£684.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611329
|
Novus Biologicals
NBP3-07918 |
50 μg |
£396.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Viperin Monoclonal antibody specifically detects Viperin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, ImmunofluorescenceSpecifications
| Viperin | |
| Western Blot, Flow Cytometry, Immunoprecipitation, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Immunology | |
| PBS containing 0.1% BSA | |
| 91543 | |
| IgG2a | |
| Protein G purified |
| MaP.VIP | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 2510004L01Rik, cig33, cig5, Cytomegalovirus-induced gene 5 protein, interferon-inducible, radical S-adenosyl methionine domain containing 2, radical S-adenosyl methionine domain-containing protein 2, vig1, Viperin, Virus inhibitory protein, endoplasmic reticulum-associated | |
| Recombinant mouse viperin fragment (residues 92-362) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title