Learn More
Thermo Scientific™ Viper™ Online SPE Solutions Kits
Ensure a perfect connection between your online SPE and your HPLC/UHPLC system with a single kit with zero-dead-volume fittings and capillaries.
Specifications
Diameter (Metric) Outer 180 µm
Type Online SPE Solutions Kit
View More Specs
Description
Description
Ensure a perfect connection between your online SPE and your HPLC/UHPLC system with the Thermo Scientific™ Dionex™ Viper™ Online SPE Solution Kits. A single kit contains Viper zero-dead-volume fittings and capillaries you need to work with your x2 Dual system. The kit contains Viper Fingertight Fittings that seal at the tip of the tubing and eliminate dead volumes and make it easier to plumb your HPLC or UHPLC system.
Conventional stainless steel HPLC connectors use a ferrule and a nut to establish a connection. A flaw of this design is that a void volume-free connection is not guaranteed, particularly when changing the tubing between differently shaped threads (e.g. when changing a column).
  • Viper connectors are standard for Dionex RSLC system connections
  • Compatible with virtually any third party UHPLC or HPLC instrument, valve, and column hardware
Specifications
Specifications
180 µm
Online SPE Solutions Kit