missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VIPAS39 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570335
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
This gene encodes a protein involved in the sorting of lysosomal proteins. Mutations in this gene are associated with ARCS2 (arthrogryposis, renal dysfunction, and cholestasis-2). Alternative splicing results in multiple transcript variants.
Specifications
| VIPAS39 | |
| Polyclonal | |
| Unconjugated | |
| VIPAS39 | |
| 6720456H09Rik; 9330175H22Rik; AI413782; C10H14orf133; C14orf133; C8H14orf133; hSPE-39; protein spe-39 homolog; SPE39; SPE-39; SPE-39 protein; Spermatogenesis-defective protein 39 homolog; VIPAR; VIPAS39; VPS16B; VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog; VPS33B-interacting protein in apical-basolateral polarity regulator; VPS33B-interacting protein in polarity and apical restriction; VPS33B-interacting protein involved in polarity and apical protein restriction | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 681989 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5PQN6 | |
| VIPAS39 | |
| synthetic peptide directed towards the middle region of rat VIPAS39. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction