missing translation for 'onlineSavingsMsg'
Learn More

VIPAR Antibody, DyLight 650, Novus Biologicals™

Product Code. 18431506 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431506 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431506 Supplier Novus Biologicals Supplier No. NBP204091C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VIPAR Polyclonal antibody specifically detects VIPAR in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen VIPAR
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias C14orf133FLJ12707, hSPE-39, Protein spe-39 homolog, SPE-39, SPE39chromosome 14 open reading frame 133, VPS16B, VPS33B interacting protein, apical-basolateral polarity regulator, VPS33B-interacting protein, VPS33B-interacting protein involved in polarity and apical protein restriction
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 443 and 493 of human VPS33B Interacting Protein, Apical-Basolateral Polarity Regulator using the numbering given in entry NP_071350.2 (GeneID 63894).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63894
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.