missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VIPAR Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-04091DL594
This item is not returnable.
View return policy
Description
VIPAR Polyclonal antibody specifically detects VIPAR in Human, Mouse samples. It is validated for Western Blot
Specifications
| VIPAR | |
| Polyclonal | |
| Western Blot | |
| C14orf133FLJ12707, hSPE-39, Protein spe-39 homolog, SPE-39, SPE39chromosome 14 open reading frame 133, VPS16B, VPS33B interacting protein, apical-basolateral polarity regulator, VPS33B-interacting protein, VPS33B-interacting protein involved in polarity and apical protein restriction | |
| The immunogen recognized by this antibody maps to a region between residue 443 and 493 of human VPS33B Interacting Protein, Apical-Basolateral Polarity Regulator using the numbering given in entry NP_071350.2 (GeneID 63894). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 63894 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction