missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VIP36 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552027
This item is not returnable.
View return policy
Description
Immunogen sequence: GNSEHLKREH SLIKPYQGVG SSSMPLWDFQ GSTMLTSQYV RLTPDERSKE GSIWNHQPCF LKDWEMHVHF KVHGTGKKNL HGDGIALWYT RDRLVPGPVF GSKDNFHGLA IFLDTYPNDE TTERVFPYIS VMVNNGSLSY DHSKD Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.
This gene encodes a type I transmembrane lectin that shuttles between the endoplasmic reticulum, the Golgi apparatus and the plasma membrane. The encoded protein binds high mannose type glycoproteins and may facilitate their sorting, trafficking and quality control.
Specifications
| VIP36 | |
| Polyclonal | |
| Unconjugated | |
| LMAN2 | |
| 1110003H06Rik; 1300009F09Rik; AA408240; AL023023; AU040819; C5orf8; glycoprotein GP36b; GP36B; Lectin mannose-binding 2; lectin, mannose binding 2; lectin, mannose-binding 2; Lman2; vesicular integral protein of 36 kDa; Vesicular integral-membrane protein 36; Vesicular integral-membrane protein VIP36; Vip36; VIP36 (vesicular integral-membrane protein); wu:fb16f04; zgc:158761 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10960 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q12907 | |
| LMAN2 | |
| Recombinant protein corresponding to Human VIP36. Recombinant protein control fragment (Product #RP-102045). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction