missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VIP36 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121508
This item is not returnable.
View return policy
Description
VIP36 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a type I transmembrane lectin that shuttles between the endoplasmic reticulum, the Golgi apparatus and the plasma membrane. The encoded protein binds high mannose type glycoproteins and may facilitate their sorting, trafficking and quality control.
Specifications
| VIP36 | |
| Polyclonal | |
| Unconjugated | |
| LMAN2 | |
| 1110003H06Rik; 1300009F09Rik; AA408240; AL023023; AU040819; C5orf8; glycoprotein GP36b; GP36B; Lectin mannose-binding 2; lectin, mannose binding 2; lectin, mannose-binding 2; Lman2; vesicular integral protein of 36 kDa; Vesicular integral-membrane protein 36; Vesicular integral-membrane protein VIP36; Vip36; VIP36 (vesicular integral-membrane protein); wu:fb16f04; zgc:158761 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 10960, 290994 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q12907 | |
| LMAN2 | |
| Recombinant protein Lectin, Mannose Binding 2. The antigen corresponds to amino acid range 45-322 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction