missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VIP Antibody (OTI1E8), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74848R
This item is not returnable.
View return policy
Description
VIP Monoclonal specifically detects VIP in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| VIP | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| VIP | |
| Full length recombinant protein of human VIP (NP_003372) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1E8 | |
| Western Blot, Immunohistochemistry | |
| MGC13587, PHM27, vasoactive intestinal peptide, VIP peptides | |
| Mouse | |
| Affinity Purified | |
| Cancer, Cell Cycle and Replication, GPCR, Neuroscience, Neurotransmission, Signal Transduction | |
| 7432.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido