missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VIP Antibody (2), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05163AF750
This item is not returnable.
View return policy
Description
VIP Monoclonal specifically detects VIP in Human, Rat, Porcine, Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| VIP | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| VIP | |
| Vasoactive intestinal peptide adsorbed onto aluminum hydroxide gel | |
| 0.1 mL | |
| Primary | |
| Binds to human, bovine, porcine or rat VIP, but does not bind to PHM or PACAP | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| MGC13587, PHM27, vasoactive intestinal peptide, VIP peptides | |
| Mouse | |
| Protein A or G purified | |
| Cancer, Cell Cycle and Replication, GPCR, Neuroscience, Neurotransmission, Signal Transduction | |
| 7432.0 | |
| Human, Rat, Pig, Bovine | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction