missing translation for 'onlineSavingsMsg'
Learn More

Violuric acid monohydrate, 97%

Product Code. p-7027289
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456796 10 g 10g
11466796 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456796 Supplier Thermo Scientific Alfa Aesar Supplier No. B23931.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26351-19-9 | C4H5N3O5 | 175.10 g/mol

Violuric acid monohydrate is used as a reagent for cobalt. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26351-19-9
Molecular Formula C4H5N3O5
Molecular Weight (g/mol) 175.10
MDL Number MFCD00149401
InChI Key YHAIHNZKUCGXRI-UHFFFAOYSA-N
Synonym violuric acid monohydrate, 5-hydroxyimino pyrimidine-2,4,6 1h,3h,5h-trione hydrate, 2,4,5,6 1h,3h-pyrimidinetetrone, 5-oxime, monohydrate, 5-hydroxyimino-1,3-diazinane-2,4,6-trione hydrate, alloxan-5-oxime monohydrate, c4nh33o4.h2o, 5-isonitrosobarbituric acid monohydrate, 5-hydroxyimino pyrimidine-2,4,6 1h,3h,5h-trionehydrate, 5-hydroxyimino-1,3-dihydropyrimidine-2,4,6-trione, oxamethane, violuric acid monohydrate, for spectrophotometric det. of co
PubChem CID 21197397
SMILES O.ON=C1C(=O)NC(=O)NC1=O

Specifications

Melting Point ∼245°C (decomposition)
Quantity 10 g
Beilstein 162342
Merck Index 14,10000
IUPAC Name 5-hydroxyimino-1,3-diazinane-2,4,6-trione;hydrate
Formula Weight 175.10 (157.08 Anhydrous)
Percent Purity 97%
Chemical Name or Material Violuric acid monohydrate
compliance-icons
Product Identifier
  • Violuric acid monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Violuric acid monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.