missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Violet Red Bile Glucose (VRBG) Agar (ISO) (Dehydrated)

Product Code. p-7079179
Click to view available options
Quantity:
500 g
2.5 kg
5 kg
Unit Size:
2.50kg
500g
5kg
This item is not returnable. View return policy

Product Code. 15812347

Brand: Thermo Scientific™ CM1082R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Detect and enumerate Enterobacteriaceae in food, animal feed, and environmental samples with this selective medium.

Selectively enumerate Enterobacteriaceae in food, animal feed, and environmental samples with Thermo Scientific™ Oxoid™ Violet Red Bile Glucose (VRBG) Agar (ISO) tested for performance based on ISO 21528 and ISO 11133:2014. Due to the careful addition of selective inhibitory components, neutral red, crystal violet, and bile salts, the medium provides selectivity against non-target Gram-positive bacteria.

Use Violet Red Bile Glucose (VRBG) Agar for examination of food, animal feed, and environmental samples, for the presence of indicator groups such as coliforms. As well as providing evidence of food hygiene and safety, indicator groups provide a measure of food quality and spoilage potential. One of the most common groups of bacteria employed as indicator organisms by the food industry are the coliforms, which can be regarded as a subgroup within the Enterobacteriaceae. However, in an attempt to improve assessment of food safety and quality by the food industry, there has been a move towards testing for total Enterobacteriaceae rather than limiting testing to coliforms.

In processed foods, particularly foods subjected to heat treatment, these organisms can provide a reliable indication of process failure, under-processing, or post-processing contamination.
  • Easy-to-read results: due to inclusion of neutral red indicator and crystal violet in the medium Enterobacteriaceae form purple pink colored colonies.
  • Tested for performance based on ISO 21528 and ISO 11133:2014 standards.
Mossel 1 modified Violet Red Bile Lactose Agar by adding glucose to improve recovery of all Enterobacteriaceae. Later work by Mossel and Cowell;2,3 demonstrated that lactose could be omitted, resulting in the formulation known as VRBGA.

Enterobacteriaceae rapidly ferment glucose and so reduce the pH of the medium, which produces purple/pink coloured colonies due to the inclusion of the neutral red indicator and crystal violet in the medium. These colonies are usually surrounded by purple halos of precipitated bile salts. Crystal violet and bile salts inhibit the growth of Gram-positive flora.

Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 10°C to 30°C
Description Violet Red Bile Glucose (VRBG) Agar (ISO) (Dehydrated)
Form Powder
Product Type Agar
Quantity 2.5 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.