missing translation for 'onlineSavingsMsg'
Learn More

Vinyltriphenylphosphonium bromide, 97%

Product Code. 11380174
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11380174 25 g 25g
11370174 5 g 5g
11390174 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11380174 Supplier Thermo Scientific Alfa Aesar Supplier No. L00708.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5044-52-0 | C20H18BrP | 369.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5044-52-0
Molecular Formula C20H18BrP
Molecular Weight (g/mol) 369.24
MDL Number MFCD00011807
InChI Key VRAYVWUMBAJVGH-UHFFFAOYSA-M
Synonym vinyltriphenylphosphonium bromide, triphenyl vinyl phosphonium bromide, schweizer's reagent, ethenyltriphenylphosphonium bromide, ethenyl triphenyl phosphanium bromide, ethenyltriphenylphosphanium bromide, triphenyl-vinyl-phosphonium; bromide, pubchem16334, acmc-1aw2n, vinyltriphenylphosphine bromide
PubChem CID 2723893
IUPAC Name ethenyl(triphenyl)phosphanium;bromide
SMILES [Br-].C=C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 182°C to 186°C
Quantity 25 g
Beilstein 4168860
Sensitivity Hygroscopic; Light sensitive
Formula Weight 369.25
Percent Purity 97%
Chemical Name or Material Vinyltriphenylphosphonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.