missing translation for 'onlineSavingsMsg'
Learn More

Vinyltrimethoxysilane, 98%

Product Code. 11464667
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464667 100 g 100g
11474667 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464667 Supplier Thermo Scientific Alfa Aesar Supplier No. B21176.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2768-02-7 | C5H12O3Si | 148.23 g/mol

Trimethoxyvinylsilane is used as a chemical intermediate in the production of other fine chemicals or as a monomer in the production of vinyl functional silicone polymers. The use of Trimethoxyvinylsilane in coatings is intended to improve bonding or adhesion between organic polymers and mineral surfaces, such as pigments, fillers, and glass or metal substrates. It is used in water-based decorative coating products. It is also used as a as cross-linking agent and adhesion promoter in commercially-available silicone sealant product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethoxyvinylsilane is used as a chemical intermediate in the production of other fine chemicals or as a monomer in the production of vinyl functional silicone polymers. The use of Trimethoxyvinylsilane in coatings is intended to improve bonding or adhesion between organic polymers and mineral surfaces, such as pigments, fillers, and glass or metal substrates. It is used in water-based decorative coating products. It is also used as a as cross-linking agent and adhesion promoter in commercially-available silicone sealant product.

Solubility
Solubility in water, 9.4 g/L at 20°C.

Notes
Moisture sensitive. Store away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-7-2768
PubChem CID 76004

Specifications

Boiling Point 122°C to 124°C
Melting Point <-40°C
Quantity 100 g
Molecular Formula C5H12O3Si
Linear Formula CH2=CHSi(OCH3)3
MDL Number MFCD00008605
UN Number UN1993
Beilstein 1099136
Flash Point 23°C (73°F)
Synonym vinyltrimethoxysilane, trimethoxyvinylsilane, silane, ethenyltrimethoxy, trimethoxy vinyl silane, trimethoxysilyl ethene, vinyl trimethoxy silane, silane, trimethoxyvinyl, trimethoxysilyl ethylene, vtmo, vts-m
InChI Key NKSJNEHGWDZZQF-UHFFFAOYSA-N
SMILES CO[Si](C=C)(OC)OC
IUPAC Name ethenyl(trimethoxy)silane
Molecular Weight (g/mol) 148.23
Formula Weight 148.24
Percent Purity 98%
Odor Fruit-like
Refractive Index 1.392
Sensitivity Moisture sensitive
Density 0.97
Chemical Name or Material Vinyltrimethoxysilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.