missing translation for 'onlineSavingsMsg'
Learn More

Vinyltrimethoxysilane, 98%, AcroSeal™

Product Code. 10423350 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10423350 1 L AcroSeal™ Glass Bottle 1 L
10713922 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10423350 Supplier Thermo Scientific Acros Supplier No. 433060010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2768-02-7 | C5H12O3Si | 148.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-7-2768
Molecular Formula C5H12O3Si
Molecular Weight (g/mol) 148.24
MDL Number MFCD00008605
InChI Key NKSJNEHGWDZZQF-UHFFFAOYSA-N
Synonym vinyltrimethoxysilane, trimethoxyvinylsilane, silane, ethenyltrimethoxy, trimethoxy vinyl silane, trimethoxysilyl ethene, vinyl trimethoxy silane, silane, trimethoxyvinyl, trimethoxysilyl ethylene, vtmo, vts-m
PubChem CID 76004
IUPAC Name ethenyl(trimethoxy)silane
SMILES CO[Si](C=C)(OC)OC

Specifications

Color Colorless
Density 0.9680g/mL
Boiling Point 123.0°C
Flash Point 23°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula H2C=CHSi(OCH3)3
Refractive Index 1.3915 to 1.3935
Quantity 1 L
Specific Gravity 0.968
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in most common organic solvents
Viscosity 0.6 mm2/s (25°C)
Formula Weight 148.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Vinyltrimethoxysilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Vinyltrimethoxysilane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Skin sensitiser Category 1
Hazard Statement
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.