missing translation for 'onlineSavingsMsg'
Learn More

Vinyltriethoxysilane, 97%

Product Code. 11490127
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490127 50 g 50g
11400137 250 g 250g
11480127 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11490127 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer B21037.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 78-08-0 | C8H18O3Si | 190.31 g/mol

Vinyltriethoxysilane is used as an adhesive for glass slides. It acts as a coupling agent on inorganic substances blended into polymers that function as adhesion promoters, crosslinkers, and surface modifiers. Further, it is used to prepare polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vinyltriethoxysilane is used as an adhesive for glass slides. It acts as a coupling agent on inorganic substances blended into polymers that function as adhesion promoters, crosslinkers, and surface modifiers. Further, it is used to prepare polymers.

Solubility
Miscible with organic solvents.

Notes
Moisture sensitive. Incompatible with alkali, acid, water and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 78-08-0
Molecular Formula C8H18O3Si
Molecular Weight (g/mol) 190.31
MDL Number MFCD00009063
InChI Key FWDBOZPQNFPOLF-UHFFFAOYSA-N
Synonym vinyltriethoxysilane, triethoxyvinylsilane, silane, ethenyltriethoxy, triethoxy vinyl silane, polyscience vtes, triethoxyvinylsilicane, vtes, silane a 151, ethenyl triethoxy silane, silane, triethoxyvinyl
PubChem CID 6516
IUPAC Name ethenyl(triethoxy)silane
SMILES CCO[Si](OCC)(OCC)C=C

Specifikationer

Density 0.903
Boiling Point 160°C to 161°C
Flash Point 34°C (93°F)
Assay Percent Range 0.97
Linear Formula (CH3CH2O)3SiCH=CH2
Refractive Index 1.399
Quantity 50 g
UN Number UN1993
Beilstein 1767229
Sensitivity Moisture sensitive
Solubility Information Miscible with organic solvents.
Formula Weight 190.31
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Vinyltriethoxysilane
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • Vinyltriethoxysilane
Signalord
  • Warning
Riskkategorier
  • Flammable liquid Category 3
Faroangivelser
  • H226-Flammable liquid and vapour.
Skyddsangivelse
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Kompletterande information
  • MIXTURE LIST-Contain: Vinyltriethoxysilane

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.