missing translation for 'onlineSavingsMsg'
Learn More

Vinyltriethoxysilane, 97%, Thermo Scientific Chemicals

Catalog Number p-7025523
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
78-08-0
C8H18O3Si
190.31
MFCD00009063
FWDBOZPQNFPOLF-UHFFFAOYSA-N
vinyltriethoxysilane, triethoxyvinylsilane, silane, ethenyltriethoxy, triethoxy vinyl silane, polyscience vtes, triethoxyvinylsilicane, vtes, silane a 151, ethenyl triethoxy silane, silane, triethoxyvinyl
6516
ethenyl(triethoxy)silane
CCO[Si](OCC)(OCC)C=C
This item is not returnable. View return policy
78-08-0
C8H18O3Si
190.31
MFCD00009063
FWDBOZPQNFPOLF-UHFFFAOYSA-N
vinyltriethoxysilane, triethoxyvinylsilane, silane, ethenyltriethoxy, triethoxy vinyl silane, polyscience vtes, triethoxyvinylsilicane, vtes, silane a 151, ethenyl triethoxy silane, silane, triethoxyvinyl
6516
ethenyl(triethoxy)silane
CCO[Si](OCC)(OCC)C=C

Vinyltriethoxysilane is used as an adhesive for glass slides. It acts as a coupling agent on inorganic substances blended into polymers that function as adhesion promoters, crosslinkers, and surface modifiers. Further, it is used to prepare polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vinyltriethoxysilane is used as an adhesive for glass slides. It acts as a coupling agent on inorganic substances blended into polymers that function as adhesion promoters, crosslinkers, and surface modifiers. Further, it is used to prepare polymers.

Solubility
Miscible with organic solvents.

Notes
Moisture sensitive. Incompatible with alkali, acid, water and strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 0.903
Boiling Point 160°C to 161°C
Flash Point 34°C (93°F)
Assay Percent Range 0.97
Linear Formula (CH3CH2O)3SiCH=CH2
Refractive Index 1.399
Quantity 50 g
UN Number UN1993
Beilstein 1767229
Sensitivity Moisture sensitive
Solubility Information Miscible with organic solvents.
Formula Weight 190.31
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Vinyltriethoxysilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'