missing translation for 'onlineSavingsMsg'
Learn More

Vinyloxytrimethylsilane, 97%

Product Code. 11397059
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397059 5 g 5g
11387059 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397059 Supplier Thermo Scientific Alfa Aesar Supplier No. H26876.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6213-94-1 | C5H12OSi | 116.235 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6213-94-1
Molecular Formula C5H12OSi
Molecular Weight (g/mol) 116.235
MDL Number MFCD00054764
InChI Key HFTNNOZFRQLFQB-UHFFFAOYSA-N
Synonym trimethyl vinyloxy silane, vinyloxytrimethylsilane, silane, ethenyloxy trimethyl, tmso-ethene, vinyloxy-trimethylsilane, trimethylsiloxy ethylene, snqhhgciiezjp@, acmc-209mzx, ethenoxy trimethyl silane, trimethylsilyl vinyl ether
PubChem CID 80344
IUPAC Name ethenoxy(trimethyl)silane
SMILES C[Si](C)(C)OC=C

Specifications

Boiling Point 74°C
Quantity 5 g
UN Number UN1993
Flash Point −19°C (−2°F)
Formula Weight 116.24
Percent Purity 97%
Refractive Index 1.389
Sensitivity Moisture sensitive
Density 0.779
Chemical Name or Material Vinyloxytrimethylsilane
compliance-icons compliance-icons
Product Identifier
  • Vinyloxytrimethylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: (ethenyloxy)trimethylsilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.