missing translation for 'onlineSavingsMsg'
Learn More

Vinylmagnesium chloride, 1.5M (13 wt.%) solution in THF, AcroSeal™

Product Code. 10043260 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10043260 800 mL AcroSeal™ Glass Bottle 800mL
10507763 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10043260 Supplier Thermo Scientific Acros Supplier No. 252598000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 3536-96-7 | C2H3ClMg | 86.80 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3536-96-7 , 109-99-9
Molecular Formula C2H3ClMg
Molecular Weight (g/mol) 86.80
MDL Number MFCD00013594
InChI Key NAFZXKWEUWVSAW-UHFFFAOYSA-M
Synonym vinylmagnesium chloride, chlorovinylmagnesium, chloro ethenyl magnesium, vinyl magnesium chloride, magnesium, chloroethenyl, vinyl magnesium-chloride, vinyl-magnesium chloride, magnesium,chloroethenyl, ethenyl magnesium chloride, vinylmagnesium chloride solution
PubChem CID 77075
IUPAC Name magnesium;ethene;chloride
SMILES Cl[Mg]C=C

Specifications

Color White to Beige
Boiling Point 66.0°C
Linear Formula CH2=CHMgCl
Density 0.9800g/mL
Solubility Information Solubility in water: vigorous reaction.
Formula Weight 86.8
Percent Purity 2M (18 wt.%)
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.98
Quantity 800 mL
Flash Point −017°C
Physical Form Crystalline Powder
Chemical Name or Material Vinylmagnesium chloride, 15 wt. % Solution in Tetrahydrofuran
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Vinylmagnesium chloride, 1.9M (16.5 wt.%) solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.