missing translation for 'onlineSavingsMsg'
Learn More

Vinylene carbonate, 98%, stabilized, Thermo Scientific Chemicals

Product Code. 10611073
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-36-6
Molecular Formula C3H2O3
Molecular Weight (g/mol) 86.05
MDL Number MFCD00005380
InChI Key VAYTZRYEBVHVLE-UHFFFAOYSA-N
Synonym vinylene carbonate, vinyl carbonate, carbonic acid, cyclic vinylene ester, wln: t5ovoj, vinylenecarbonate, vinyleny carbonate, carbonic acid vinylene, 1,3-dioxo-2-one, vinylene carbonate vc, acmc-209qj3
PubChem CID 13385
SMILES O=C1OC=CO1

Specifications

Melting Point 19°C to 22°C
Color Colorless to Yellow
Density 1.3600g/mL
Boiling Point 162°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4200 to 1.423
Quantity 25 g
Beilstein 19, V,4, 72
Fieser 13,342
Specific Gravity 1.36
Solubility Information Solubility in water: 11.5g / 100mL
IUPAC Name 1,3-dioxol-2-one
Stabilizer 0.1% max. BHT
Formula Weight 86.05
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Vinylene carbonate, Stabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Vinylene carbonate, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H317-May cause an allergic skin reaction.,May cause an allergic skin reaction.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.,Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap and water.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/physician.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'