missing translation for 'onlineSavingsMsg'
Learn More

Vinylene carbonate, 97+%, stab. with BHT, Thermo Scientific Chemicals

Product Code. 11354108
Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Vinylene carbonate is used as an additive to electrolyte solutions for anode side Lithium-ion batteries. It also acts as a sealant to seal at least a portion of the silicon-polyvinyl acid interface. It is further used for great improvement of high temperature performance of the battery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-36-6
Molecular Formula C3H2O3
Molecular Weight (g/mol) 86.05
MDL Number MFCD00005380
InChI Key VAYTZRYEBVHVLE-UHFFFAOYSA-N
Synonym vinylene carbonate, vinyl carbonate, carbonic acid, cyclic vinylene ester, wln: t5ovoj, vinylenecarbonate, vinyleny carbonate, carbonic acid vinylene, 1,3-dioxo-2-one, vinylene carbonate vc, acmc-209qj3
PubChem CID 13385
IUPAC Name 1,3-dioxol-2-one
SMILES O=C1OC=CO1

Specifications

Melting Point 19°C to 22°C
Color Colorless to Yellow
Density 1.355 g/mL
Boiling Point 162°C to 163°C
Flash Point 72°C (163°F)
Refractive Index 1.421
Quantity 0.5 g
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 86.05
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material Vinylene carbonate, Stabilized with BHT
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'