missing translation for 'onlineSavingsMsg'
Learn More

Vinylene carbonate, 97+%, stab. with BHT

Product Code. 11334108
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334108 2 g 2g
11354108 0.5 g 0.5g
11344108 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334108 Supplier Thermo Scientific Alfa Aesar Supplier No. 041400.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 872-36-6 | C3H2O3 | 86.05 g/mol

Vinylene carbonate is used as an additive to electrolyte solutions for anode side Lithium-ion batteries. It also acts as a sealant to seal at least a portion of the silicon-polyvinyl acid interface. It is further used for great improvement of high temperature performance of the battery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-36-6
Molecular Formula C3H2O3
Molecular Weight (g/mol) 86.05
MDL Number MFCD00005380
InChI Key VAYTZRYEBVHVLE-UHFFFAOYSA-N
Synonym vinylene carbonate, vinyl carbonate, carbonic acid, cyclic vinylene ester, wln: t5ovoj, vinylenecarbonate, vinyleny carbonate, carbonic acid vinylene, 1,3-dioxo-2-one, vinylene carbonate vc, acmc-209qj3
PubChem CID 13385
IUPAC Name 1,3-dioxol-2-one
SMILES O=C1OC=CO1

Specifications

Melting Point 19°C to 22°C
Color Colorless to Yellow
Density 1.355 g/mL
Boiling Point 162°C to 163°C
Flash Point 72°C (163°F)
Refractive Index 1.421
Quantity 2 g
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 86.05
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material Vinylene carbonate, Stabilized with BHT
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.