missing translation for 'onlineSavingsMsg'
Learn More

Vinylcyclohexane (Stabilized by TBC), TRC

Product Code. 30133968
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133968 2.5 g 2.5g
30136843 500 mg 500mg
30012353 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133968 Supplier Toronto Research Chemicals Supplier No. TRCV7570802.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 695-12-5
Molecular Formula C8H14
Molecular Weight (g/mol) 110.2
IUPAC Name ethenylcyclohexane
SMILES C=CC1CCCCC1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C8H14/c1-2-8-6-4-3-5-7-8/h2,8H,1,3-7H2
Formula Weight 110.1096
Chemical Name or Material Vinylcyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.