missing translation for 'onlineSavingsMsg'
Learn More

Vinylcyclohexane, 97%

Product Code. p-7045446
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11372976

Brand: Thermo Scientific Alfa Aesar L10756.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 695-12-5 | C8H14 | 110.20 g/mol

The use of vinylcyclohexane dioxide (VCD) as a polar dehydrant with subsequent embedment in Spurr was studied. Vinylcyclohexane (VCH) was copolymerized with ethene and propene using methylaluminoxane-activated metallocene catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The use of vinylcyclohexane dioxide (VCD) as a polar dehydrant with subsequent embedment in Spurr was studied. Vinylcyclohexane (VCH) was copolymerized with ethene and propene using methylaluminoxane-activated metallocene catalysts.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 695-12-5
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00001514
InChI Key LDLDYFCCDKENPD-UHFFFAOYSA-N
Synonym vinylcyclohexane, cyclohexylethylene, cyclohexane, ethenyl, cyclohexane, vinyl, cyclohexylethene, unii-q49pj3tt00, cyclohexane, ethenyl-9ci, vinyl cyclohexane, vinyl-cyclohexane, 1-vinylcyclohexane
PubChem CID 12757
IUPAC Name ethenylcyclohexane
SMILES C=CC1CCCCC1

Specifications

Density 0.805
Boiling Point 127°C to 128°C
Flash Point 22°C (71°F)
Refractive Index 1.4465
Quantity 5 g
UN Number UN3295
Beilstein 1304663
Solubility Information Insoluble in water.
Formula Weight 110.2
Percent Purity 97%
Chemical Name or Material Vinylcyclohexane
Show More Show Less
compliance-icons
Product Identifier
  • Vinylcyclohexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Supplemental information
  • MIXTURE LIST-Contains : Vinylcyclohexane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.