missing translation for 'onlineSavingsMsg'
Learn More

Vinylacetic Acid, TRC

Product Code. 30449508
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449508 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449508 Supplier Toronto Research Chemicals Supplier No. TRCV42400025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vinylacetic Acid
CAS 625-38-7
Molecular Formula C4H6O2
Quantity 25 g
Synonym 3-Butenoic Acid, Ethenylacetic Acid, NSC 44546, β-Butenoic Acid
InChI Formula InChI=1S/C4H6O2/c1-2-3-4(5)6/h2H,1,3H2,(H,5,6)
SMILES OC(CC=C)=O
IUPAC Name but-3-enoic acid
Molecular Weight (g/mol) 86.09
Formula Weight 86.0368
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.