missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Vinylacetic acid, tech., 90%, unstabilized

Product Code. 10377330 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10377330 25 g Glass bottle 25g
10543901 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10377330 Supplier Thermo Scientific Chemicals Supplier No. 158830250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 625-38-7 | C4H6O2 | 86.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-38-7
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00002782
InChI Key PVEOYINWKBTPIZ-UHFFFAOYSA-N
Synonym 3-butenoic acid, vinylacetic acid, vinyl acetic acid, acetic acid, ethenyl, beta-butenoic acid, .beta.-butenoic acid, 2-propenylcarboxylic acid, allylic acid, vinylessigsaeure, 3-butenoicacid
PubChem CID 32743
ChEBI CHEBI:35897
IUPAC Name but-3-enoic acid
SMILES OC(=O)CC=C

Specifications

Melting Point -39°C
Color Colorless to Yellow
Density 1.0100g/mL
Boiling Point 163°C
Flash Point 65°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula H2C=CHCH2CO2H
Refractive Index 1.4239 to 1.4259
Quantity 25 g
Beilstein 02, 407
Specific Gravity 1.01
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most common organic solvents
Formula Weight 86.09
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Vinylacetic acid, tech., 90%, unstabilized
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.