missing translation for 'onlineSavingsMsg'
Learn More

Vinylacetic acid, 96%

Product Code. 11341947
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341947 100 g 100g
11331947 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11341947 Supplier Thermo Scientific Alfa Aesar Supplier No. L07557.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 625-38-7 | C4H6O2 | 86.09 g/mol

Vinylacetic acid used in the preparation of strained nitroso Diels-Aldrer adducts which undergo a domino ROC metathesis in the presence of Ru-carbene catalysts and olefins. It is used in the synthesis of polyethylene glycolated boltorn coatings. It was used to study the inhibition of the enzyme-catalyzed conversion of D-tyrosyl- phenylalanyl-glycine to D-tyrosyl-phenylaline amide by non-peptide carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vinylacetic acid used in the preparation of strained nitroso Diels-Aldrer adducts which undergo a domino ROC metathesis in the presence of Ru-carbene catalysts and olefins. It is used in the synthesis of polyethylene glycolated boltorn coatings. It was used to study the inhibition of the enzyme-catalyzed conversion of D-tyrosyl- phenylalanyl-glycine to D-tyrosyl-phenylaline amide by non-peptide carboxylic acids.

Solubility
Fully miscible.

Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents. Incompatible with oxidizing agents, heat, Bases, Reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-38-7
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00002782
InChI Key PVEOYINWKBTPIZ-UHFFFAOYSA-N
Synonym 3-butenoic acid, vinylacetic acid, vinyl acetic acid, acetic acid, ethenyl, beta-butenoic acid, .beta.-butenoic acid, 2-propenylcarboxylic acid, allylic acid, vinylessigsaeure, 3-butenoicacid
PubChem CID 32743
ChEBI CHEBI:35897
IUPAC Name but-3-enoic acid
SMILES OC(=O)CC=C

Specifications

Melting Point -39°C
Density 1.012
Boiling Point 162°C to 163°C
Flash Point 65°C (149°F)
Linear Formula H2C=CHCH2CO2H
Refractive Index 1.424
Quantity 100 g
UN Number UN3265
Beilstein 1699159
Solubility Information Fully miscible.
Formula Weight 86.09
Percent Purity 96%
Chemical Name or Material Vinylacetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.