missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Vinyl Safety
Vinyl Safety
Specifications
Specifications
| Color | Red, White |
| Material | Laminated Vinyl |
| Product Type | Safety Tape |
| For Use With (Application) | Marking equipment/exits, hazardous areas and first aid points |
| Size (Metric) | 16 m x 51 mm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?