missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Vinyl Printer Tape

Product Code. 11574044
Change view
Click to view available options
Color:
White
Length (Metric):
15.24 m
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Length (Metric) unitSize
11574044 White 15.24 m Each
11583924 White 15.24 m Each
11563974 White 15.24 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11574044 Supplier Brady™ Supplier No. 114835

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Description BMP71 Label Printer Tapes
Adhesive Type Acrylic, Pressure-sensitive
Color White
Finish Gloss
Material Vinyl
For Use With (Application) Security identification
For Use With (Equipment) BMP™71
Gloss 60° 70 Gardner Units
Labels per Row 1
Quantity 1 Ea.
Temperature Range (Metric) -40°C to +90°C
Thickness (Metric) 0.015 mm
Length (Metric) 15.24 m
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.