missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Vinyl Pre-Printed Chemical Labels

Product Code. 13573340
Change view
Click to view available options
Description:
1 Diamond
2 Diamonds, vertical
3 Diamonds
Height (Metric):
155 mm
155.00 mm
Width (Metric):
105 mm
Unit Size:
1 roll
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Height (Metric) Width (Metric) unitSize
13573340 2 Diamonds, vertical 155.00 mm 105 mm 1 roll
15326651 3 Diamonds 155 mm 105 mm 1 roll
13543340 1 Diamond 155.00 mm 105 mm 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13573340 Supplier Brady™ Supplier No. 121646
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Brady™ Vinyl GHS/CLP Chemical Labels for BBP3x printers are self-adhesive and preprinted with red diamonds in a variety of layouts.

  • BBP™31 and BBP™33 compatible labels
  • Blank labels with pre-printed red diamonds allow printer to produce two-color GHS/CLP labels in one print pass
  • Available in three sizes in the full product portfolio, each with six diamond layouts that match templates in MarkWare™Software
  • Service temperature -20°C to 75°C; minimum application temperature 10°C
  • Average outdoor durability 3-5 years

Specifications

Certifications/Compliance BS5609:1986 Section 2 and section 3
Description 2 Diamonds, vertical
Adhesive Type Acrylic, Permanent
Color White, Red
Finish Gloss
Material Vinyl
Product Type Pre-printed Chemical Label
For Use With (Application) Facility and Safety identification, Hazardous Substance identification
Labels per Row 1
Life Span Up to 3 Years Outdoor Use
Quantity 200/Pk.
Software Type MarkWare
Temperature Range (Metric) -20°C to +75°C
Thickness (Metric) 0.026 mm
Dimensions (L x W) 155 x 105 mm
Width (Metric) 105 mm
Height (Metric) 155.00 mm
Height (Metric) Character 155 mm
Weight (Metric) 0.1 kg
Alternate Part Number B30-262-7569-CLP2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.